MilliporeSigma
  • Curcumin nanoparticle doped starch thin film as a green colorimetric sensor for detection of boron.
Curcumin nanoparticle doped starch thin film as a green colorimetric sensor for detection of boron.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2019-07-25)
Chanita Boonkanon, Kharittha Phatthanawiwat, Worawit Wongniramaikul, Aree Choodum
ABSTRACT
A tapioca starch film doped with curcumin nanoparticles was successfully fabricated and applied as a novel green colorimetric sensor for detection of boron in wastewater. Curcumin nanoparticles (curn, 30-90 nm) extracted from turmeric powder were used as a green probe, while tapioca starch was used as a natural support substrate. A yellow thin film (51 μm thick) fabricated on a used plastic spoon turned red-brown after immersion in boron solution (pH 9) for 15 min with excellent selectivity. The film costs only 0.0007 USD, while the cost of the sensor (curn-film on new plastic spoon) was 0.004 USD. After use the film could be completely washed from the plastic, it being biodegradable, while the used plastic spoon could be re-used to fabricate a new sensor at least 10 times. The good 1.52%RSD precision was obtained across three lots fabricated. When the curn-film was used in conjunction with digital image colorimetry (DIC), a simple and rapid quantification of boron was achieved. The green color layer in reflected light image of the red-brown product (IG) provided the highest sensitivity (64 ± 1 a.u. L mg-1) and the lowest detection limit of 0.052 ± 0.001 mg L-1. The intra-day testing (9 films) had 2.41 to 4.34%RSD, while the inter-day testing had 2.29 to 5.66%RSD (15 films, 5 days). Accuracy in terms of relative error for control samples (0.40 mg L-1) was +3.63%. Wastewater samples from Para-rubber wood processing plant were quantified by curn-film and DIC, giving 4248 ± 391 mg L-1 boron concentration with no significant difference to ICP determination at 95% confidence level. The sensors after storage in a desiccator for a year gave readings changed by only +3.5% and -2.1% relative to freshly prepared sensors.
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Boron, monofilament, 0.080 mm diameter, length 100 m