MilliporeSigma
  • Citrullinated Peptide Epitope Targets Therapeutic Nanoparticles to Human Neutrophils.
Citrullinated Peptide Epitope Targets Therapeutic Nanoparticles to Human Neutrophils.
Bioconjugate chemistry (2019-09-17)
Sangita Khatri, Jonas Hansen, Ana C Mendes, Ioannis S Chronakis, Shu-Chen Hung, Elizabeth D Mellins, Kira Astakhova
ABSTRACT
Multiple drugs have been proposed for reducing harsh symptoms of human rheumatic diseases. However, a targeted therapy with mild to no side effects is still missing. In this study, we have prepared and tested a series of therapeutic nanoparticles for specific targeting of human neutrophils associated with rheumatoid arthritis. In doing this, a series of citrullinated peptide epitopes derived from human proteins, fibrinogen, vimentin, and histone 3, were screened with regard to specific recognition of neutrophils. The most potent epitope proved to be a mutated fragment of an alpha chain in human fibrinogen. Next, a straightforward synthetic strategy was developed for nanoparticles decorated with this citrullinated peptide epitope and an antisense oligonucleotide targeting disease associated microRNA miR-125b-5p. Our study shows that the nanoparticles specifically recognize neutrophils and knock down miR-125b-5p, with no apparent toxicity to human cells. In contrast to organic dendrimers, chitosan-hyaluronic acid formulations do not activate human innate immune response. Our data proves that the strategy we report herein is effective in developing peptide epitopes for decorating delivery vehicles bearing biological drugs, targeted to a specific cell type.
Sigma-Aldrich
2,4-Dihydroxybenzophenone, 99%
Sigma-Aldrich
Mammalian Cell Lysis Kit