MilliporeSigma
  • The Role of TTP Phosphorylation in the Regulation of Inflammatory Cytokine Production by MK2/3.
The Role of TTP Phosphorylation in the Regulation of Inflammatory Cytokine Production by MK2/3.
Journal of immunology (Baltimore, Md. : 1950) (2019-09-19)
Natalia Ronkina, Nelli Shushakova, Christopher Tiedje, Tatiana Yakovleva, Maxim A X Tollenaere, Aaron Scott, Tanveer Singh Batth, Jesper Velgaard Olsen, Alexandra Helmke, Simon Holst Bekker-Jensen, Andrew R Clark, Alexey Kotlyarov, Matthias Gaestel
ABSTRACT
Tristetraprolin (TTP) is an RNA-binding protein and an essential factor of posttranscriptional repression of cytokine biosynthesis in macrophages. Its activity is temporally inhibited by LPS-induced p38MAPK/MAPKAPK2/3-mediated phosphorylation, leading to a rapid increase in cytokine expression. We compared TTP expression and cytokine production in mouse bone marrow-derived macrophages of different genotypes: wild type, MAPKAP kinase 2 (MK2) deletion (MK2 knockout [KO]), MK2/3 double deletion (MK2/3 double KO [DKO]), TTP-S52A-S178A (TTPaa) knock-in, as well as combined MK2 KO/TTPaa and MK2/3 DKO/TTPaa. The comparisons reveal that MK2/3 are the only LPS-induced kinases for S52 and S178 of TTP and the role of MK2 and MK3 in the regulation of TNF biosynthesis is not restricted to phosphorylation of TTP at S52/S178 but includes independent processes, which could involve other TTP phosphorylations (such as S316) or other substrates of MK2/3 or p38MAPK Furthermore, we found differences in the dependence of various cytokines on the cooperation between MK2/3 deletion and TTP mutation ex vivo. In the cecal ligation and puncture model of systemic inflammation, a dramatic decrease of cytokine production in MK2/3 DKO, TTPaa, and DKO/TTPaa mice compared with wild-type animals is observed, thus confirming the role of the MK2/3/TTP signaling axis in cytokine production also in vivo. These findings improve our understanding of this signaling axis and could be of future relevance in the treatment of inflammation.
Sigma-Aldrich
Thymidine 5′-triphosphate sodium salt, ≥96%
Sigma-Aldrich
PF-3644022 hydrate, ≥98% (HPLC)
Sigma-Aldrich
Cell Dissociation Solution Non-enzymatic 1x, Prepared in Hanks′ Balanced Salt Solution without calcium and magnesium, sterile-filtered, BioReagent, suitable for cell culture