MilliporeSigma
  • Biophysical studies on the antimicrobial activity of linearized esculentin 2EM.
Biophysical studies on the antimicrobial activity of linearized esculentin 2EM.
Biochimica et biophysica acta. Biomembranes (2019-12-04)
Erum Malik, David A Phoenix, Kamal Badiani, Timothy J Snape, Frederick Harris, Jaipaul Singh, Leslie Hugh Glyn Morton, Sarah R Dennison
ABSTRACT
Linearized esculentin 2 EM (E2EM-lin) from the frog, Glandirana emeljanovi was highly active against Gram-positive bacteria (minimum lethal concentration ≤ 5.0 μM) and strongly α-helical in the presence of lipid mimics of their membranes (>55.0%). The N-terminal α-helical structure adopted by E2EM-lin showed the potential to form a membrane interactive, tilted peptide with an hydrophobicity gradient over residues 9 to 23. E2EM-lin inserted strongly into lipid mimics of membranes from Gram-positive bacteria (maximal surface pressure changes ≥5.5 mN m-1), inducing increased rigidity (Cs-1 ↑), thermodynamic instability (ΔGmix < 0 → ΔGmix > 0) and high levels of lysis (>50.0%). These effects appeared to be driven by the high anionic lipid content of membranes from Gram-positive bacteria; namely phosphatidylglycerol (PG) and cardiolipin (CL) species. The high levels of α-helicity (60.0%), interaction (maximal surface pressure change = 6.7 mN m-1) and lysis (66.0%) shown by E2EM-lin with PG species was a major driver in the ability of the peptide to lyse and kill Gram-positive bacteria. E2EM-lin also showed high levels of α-helicity (62.0%) with CL species but only low levels of interaction (maximal surface pressure change = 2.9 mN m-1) and lysis (21.0%) with the lipid. These combined data suggest that E2EM-lin has a specificity for killing Gram-positive bacteria that involves the formation of tilted structure and appears to be primarily driven by PG-mediated membranolysis. These structure/function relationships are used to help explain the pore forming process proposed to describe the membranolytic, antibacterial action of E2EM-lin.
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Avanti
14:0 PE, Avanti Research - A Croda Brand
Avanti
14:0 Cardiolipin (sodium salt), 1′,3′-bis[1,2-dimyristoyl-sn-glycero-3-phospho]-glycerol (sodium salt), powder
Avanti
14:0 Cardiolipin (sodium salt), 1′,3′-bis[1,2-dimyristoyl-sn-glycero-3-phospho]-glycerol (sodium salt), chloroform
Avanti
14:0 PG, Avanti Research - A Croda Brand