MilliporeSigma
  • Analytical and ecotoxicological studies on degradation of fluoxetine and fluvoxamine by potassium ferrate.
Analytical and ecotoxicological studies on degradation of fluoxetine and fluvoxamine by potassium ferrate.
Environmental technology (2018-05-15)
Przemysław Drzewicz, Agata Drobniewska, Katarzyna Sikorska, Grzegorz Nałęcz-Jawecki
ABSTRACT
A large amount of pharmaceuticals are flushed to environment via sewage system. The compounds are persistent in environment and are very difficult to remove in drinking water treatment processes. Degradation of fluoxetine (FLU) and fluvoxamine (FLX) by ferrate(VI) were investigated. For the 10 mg/L of FLU and FLX, 35% and 50% of the compounds were degraded in the presence of 50 mg/L FeO42- within 10 minutes, respectively. After 10 minutes of the reaction, degradation of FLU and FLX is affected by formation of by-products which were likely more reactive with ferrate and competed in the reaction with FeO42-. In the case of FLU, the identified degradation by-products were hydrofluoxetine, N-methyl-3-phenyl-2-propen-1-amine, 4-(trifluoromethyl)phenol and 1-{[(1R,S)-1-Phenyl-2-propen-1-yl]oxy}-4-(trifluoromethyl)benzene. In the case of FLX, the degradation by-products were fluvoxamine acid and 5-methoxy-1-[4-(trifluoromethyl)phenyl]pent-2-en-1-imine. The results of the ecotoxicological study based on protozoa Spirostomum ambiguum have shown that 50 mg/L FeO42- reduced toxicity of 10 mg/L of FLU and FLX by around 50%. However, in the case of FLX, the results of the ecotoxicological study suggested formation of slightly more toxic compound(s) than FLX during reaction with FeO42-. Application of ferrate(VI) is a viable option for drinking water treatment process; however, caution is needed due to formation of by-products with unknown human health risk.
Sigma-Aldrich
4-(Trifluoromethyl)phenol, 97%