MilliporeSigma
  • Defects in t6A tRNA modification due to GON7 and YRDC mutations lead to Galloway-Mowat syndrome.
Defects in t6A tRNA modification due to GON7 and YRDC mutations lead to Galloway-Mowat syndrome.
Nature communications (2019-09-05)
Christelle Arrondel, Sophia Missoury, Rozemarijn Snoek, Julie Patat, Giulia Menara, Bruno Collinet, Dominique Liger, Dominique Durand, Olivier Gribouval, Olivia Boyer, Laurine Buscara, Gaëlle Martin, Eduardo Machuca, Fabien Nevo, Ewen Lescop, Daniela A Braun, Anne-Claire Boschat, Sylvia Sanquer, Ida Chiara Guerrera, Patrick Revy, Mélanie Parisot, Cécile Masson, Nathalie Boddaert, Marina Charbit, Stéphane Decramer, Robert Novo, Marie-Alice Macher, Bruno Ranchin, Justine Bacchetta, Audrey Laurent, Sophie Collardeau-Frachon, Albertien M van Eerde, Friedhelm Hildebrandt, Daniella Magen, Corinne Antignac, Herman van Tilbeurgh, Géraldine Mollet
ABSTRACT
N6-threonyl-carbamoylation of adenosine 37 of ANN-type tRNAs (t6A) is a universal modification essential for translational accuracy and efficiency. The t6A pathway uses two sequentially acting enzymes, YRDC and OSGEP, the latter being a subunit of the multiprotein KEOPS complex. We recently identified mutations in genes encoding four out of the five KEOPS subunits in children with Galloway-Mowat syndrome (GAMOS), a clinically heterogeneous autosomal recessive disease characterized by early-onset steroid-resistant nephrotic syndrome and microcephaly. Here we show that mutations in YRDC cause an extremely severe form of GAMOS whereas mutations in GON7, encoding the fifth KEOPS subunit, lead to a milder form of the disease. The crystal structure of the GON7/LAGE3/OSGEP subcomplex shows that the intrinsically disordered GON7 protein becomes partially structured upon binding to LAGE3. The structure and cellular characterization of GON7 suggest its involvement in the cellular stability and quaternary arrangement of the KEOPS complex.
Sigma-Aldrich
Nuclease P1 from Penicillium citrinum, lyophilized powder, ≥200 units/mg protein (E1%/280, 3′-5′-Phosphodiesterase)
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-TPRKB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Phosphodiesterase I from Crotalus adamanteus venom, vial of ≥100 units, Purified
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse