MilliporeSigma
  • Effects of prostaglandin F2α on angiogenic and steroidogenic pathways in the bovine corpus luteum may depend on its route of administration.
Effects of prostaglandin F2α on angiogenic and steroidogenic pathways in the bovine corpus luteum may depend on its route of administration.
Journal of dairy science (2019-09-16)
A W Jonczyk, K K Piotrowska-Tomala, P Kordowitzki, D J Skarzynski
ABSTRACT
Prostaglandin (PG) F2α and its analogs (aPGF2α) are used to induce regression of the corpus luteum (CL); their administration during the middle stage of the estrous cycle causes luteolysis in cattle. However, the bovine CL is resistant to the luteolytic actions of aPGF2α in the early stage of the estrous cycle. The mechanisms underlying this differential luteal sensitivity, as well as acquisition of luteolytic sensitivity by the CL, are still not fully understood. Therefore, to characterize possible differences in response to aPGF2α administration, we aimed to determine changes in expression of genes related to (1) angiogenesis-fibroblast growth factor 2 (FGF2), fibroblast growth factor receptor 1 (FGFR1), fibroblast growth factor receptor 2 (FGFR2), vascular endothelial growth factor A (VEGFA), vascular endothelial growth factor receptor 1 (VEGFR1), vascular endothelial growth factor receptor 2 (VEGFR2); and (2) steroidogenesis-steroidogenic acute regulatory protein (STAR), cytochrome P450 family 11 subfamily A member 1 (P450scc), and hydroxy-delta-5-steroid dehydrogenase, 3 β- and steroid delta-isomerase 1 (HSD3B) in early- and middle-stage CL that accompany local (intra-CL) versus systemic (i.m.) aPGF2α injection. Cows at d 4 (early stage) or d 10 (middle stage) of the estrous cycle were treated as follows: (1) systemic saline injection, (2) systemic aPGF2α injection (25 mg), (3) local saline injection, and (4) local aPGF2α injection (2.5 mg). Progesterone (P4) concentration was measured in jugular vein blood samples during the entire set of experiments. After 4 h of treatment, CL were collected by ovariectomy, and mRNA and protein expression levels were determined by reverse transcription quantitative-PCR and Western blotting, respectively. Local and systemic aPGF2α injections upregulated FGF2 expression but decreased expression of VEGFA in both CL stages. Both aPGF2α injections increased the expression of STAR in early-stage CL, but downregulated it in middle-stage CL. In the early-stage CL, local administration of aPGF2α upregulated HSD3B, whereas systemic injection decreased its mRNA expression in early- and middle-stage CL. Moreover, we observed a decrease in the P4 level earlier after local aPGF2α injection than after systemic administration. These results indicate that aPGF2α acting locally may play a luteotrophic role in early-stage CL. The systemic effect of aPGF2α on the mRNA expression of genes participating in steroidogenesis seems to be more substantial than its local effect in middle-stage CL.
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Fibroblast Growth Factor Receptor-1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Fibroblast Growth Factor-Basic (1-24) antibody produced in rabbit, fractionated antiserum, lyophilized powder
Sigma-Aldrich
Anti-Fibroblast Growth Factor Receptor-2, Extracellular antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution