MilliporeSigma
  • Preparation and Coagulation Performance of Carboxypropylated and Carboxypentylated Lignosulfonates for Dye Removal.
Preparation and Coagulation Performance of Carboxypropylated and Carboxypentylated Lignosulfonates for Dye Removal.
Biomolecules (2019-08-23)
Khatereh Bahrpaima, Pedram Fatehi
ABSTRACT
In this work, 1-carboxypropyled (1-CPRLS) and 5-carboxypentyled lignosulfonates (5-CPELS) were synthesized using 2-chlorobutanoic acid and 6-chlorohexanoic acid as carboxylate group donors via SN1 and SN2 mechanisms, respectively. 1-Carboxypropyl and 5-carboxypentyl lignosulfonates with the charge densities of -3.45 and -2.94 meq g-1 and molecular weights of 87,900 and 42,400 g·mol-1 were produced, respectively, under mild conditions. The carboxylate content and degree of substitution (DS) of the 1-CPRLS product were 2.37 mmol·g-1 and 0.70 mol·mol-1, while those of 5-CPELS products were 2.13 mmol·g-1 and 0.66 mol·mol-1, respectively. The grafting of carboxypropyl and carboxypentyl groups to lignosulfonate was confirmed by Fourier transform infrared (FT-IR) and nuclear magnetic resonance (1H-NMR and 13C-NMR) spectroscopies. In addition, 1-CPRLS and 5-CPELS were applied as coagulants for removing ethyl violet (EV) dye from a simulated solution, and their performance was related to their charge densities and molecular weights. Furthermore, fundamental discussion is provided on the advantages of (1) producing 1-CPRLS and (2) the superior properties and performance of 1-CPRLS to carboxyethylated lignosulfonate.
Product Description
Supelco
Acetic anhydride, derivatization grade (GC derivatization), LiChropur, ≥99.0%
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)