MilliporeSigma
Axoplasm isolation from peripheral nerve.
Developmental neurobiology (2009-11-04)
Ida Rishal, Izhak Michaelevski, Meir Rozenbaum, Vera Shinder, Katalin F Medzihradszky, Alma L Burlingame, Mike Fainzilber
ABSTRACT
Localized changes in the composition of axonal cytoplasm (axoplasm) are critical for many biological processes, including axon guidance, responses to injury, neurite outgrowth, and axon-glia interactions. Biochemical and molecular studies of these mechanisms have been heavily focused on in vitro systems because of the difficulty of obtaining subcellular extracts from mammalian tissues in vivo. As in vitro systems might not replicate the in vivo situation, reliable methods of axoplasm extraction from whole nerve would be helpful for mechanistic studies on axons. Here we develop and evaluate a new procedure for preparation of axoplasm from rat peripheral nerve, based on incubation of separated short segements of nerve fascicles in hypotonic medium to separate myelin and lyse nonaxonal structures, followed by extraction of the remaining axon-enriched material. We show that this new procedure reduces serum and glial cell contamination and facilitates proteomic analyses of axonal contents.
Sigma-Aldrich
Anti-CNPase Antibody, clone 11-5B, clone 11-5B, Chemicon®, from mouse
Sigma-Aldrich
Anti-Neurofilament H (200 kDa) Antibody, CT, serum, Chemicon®
Sigma-Aldrich
Anti-Dynein Antibody, 74 kDa Intermediate chains, cytoplasmic, clone 74.1, clone 74.1, Chemicon®, from mouse