MilliporeSigma
  • Metabolism of mephentermine and its derivatives by the microsomal fraction from male Wistar rat livers.
Metabolism of mephentermine and its derivatives by the microsomal fraction from male Wistar rat livers.
Xenobiotica; the fate of foreign compounds in biological systems (1993-01-01)
M A Mori, M Kobayashi, H Uemura, Y Mori, T Miyahara, H Kozuka
ABSTRACT
1. The N-demethylation of mephentermine (MP), p-hydroxymephentermine (p-hydroxy-MP) and N-hydroxymephentermine (N-hydroxy-MP), to produce phentermine (Ph), p-hydroxyphentermine (p-hydroxy-Ph) and N-hydroxyphentermine (N-hydroxy-Ph), and the p-hydroxylation of MP and Ph, to produce p-hydroxy-MP and p-hydroxy-Ph, were examined using rat liver microsomal preparations containing NADPH. Microsomal reduction of N-hydroxy-Ph to Ph was also examined using various cofactors. In addition, enzymic system for the N-demethylation and p-hydroxylation were examined using various inhibitors. 2. N-Hydroxy-MP demethylation to N-hydroxy-Ph proceeded at a rate almost 10-fold faster than other reactions. MP demethylation to Ph, MP oxidation to P-hydroxy-MP, Ph oxidation to p-hydroxy-Ph proceeded at similar rates, whilst p-hydroxy-MP demethylation to p-hydroxy-Ph was catalysed at the slowest rate. Microsomal reduction of N-hydroxy-Ph to Ph required NADH, and the activity was similar to that of MP oxidation to p-hydroxy-MP. 3. N-Demethylation of MP, p-hydroxy-MP and N-hydroxy-MP were inhibited not only by inhibitors of cytochrome P450, but also by methimazole, an inhibitor of the FAD-monooxygenase system. p-Hydroxylations of MP and Ph were inhibited only by inhibitors of cytochrome P450.