MilliporeSigma
Epigenetic histone methylation modulates fibrotic gene expression.
Journal of the American Society of Nephrology : JASN (2010-10-12)
Guangdong Sun, Marpadga A Reddy, Hang Yuan, Linda Lanting, Mitsuo Kato, Rama Natarajan
ABSTRACT
TGF-β1-induced expression of extracellular matrix (ECM) genes plays a major role in the development of chronic renal diseases such as diabetic nephropathy. Although many key transcription factors are known, mechanisms involving the nuclear chromatin that modulate ECM gene expression remain unclear. Here, we examined the role of epigenetic chromatin marks such as histone H3 lysine methylation (H3Kme) in TGF-β1-induced gene expression in rat mesangial cells under normal and high-glucose (HG) conditions. TGF-β1 increased the expression of the ECM-associated genes connective tissue growth factor, collagen-α1[Ι], and plasminogen activator inhibitor-1. Increased levels of chromatin marks associated with active genes (H3K4me1, H3K4me2, and H3K4me3), and decreased levels of repressive marks (H3K9me2 and H3K9me3) at these gene promoters accompanied these changes in expression. TGF-β1 also increased expression of the H3K4 methyltransferase SET7/9 and recruitment to these promoters. SET7/9 gene silencing with siRNAs significantly attenuated TGF-β1-induced ECM gene expression. Furthermore, a TGF-β1 antibody not only blocked HG-induced ECM gene expression but also reversed HG-induced changes in promoter H3Kme levels and SET7/9 occupancy. Taken together, these results show the functional role of epigenetic chromatin histone H3Kme in TGF-β1-mediated ECM gene expression in mesangial cells under normal and HG conditions. Pharmacologic and other therapies that reverse these modifications could have potential renoprotective effects for diabetic nephropathy.
Sigma-Aldrich
Normal Mouse IgG, Normal Mouse IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.