MilliporeSigma
  • Cell-Line Selectivity Improves the Predictive Power of Pharmacogenomic Analyses and Helps Identify NADPH as Biomarker for Ferroptosis Sensitivity.
Cell-Line Selectivity Improves the Predictive Power of Pharmacogenomic Analyses and Helps Identify NADPH as Biomarker for Ferroptosis Sensitivity.
Cell chemical biology (2016-02-09)
Kenichi Shimada, Miki Hayano, Nen C Pagano, Brent R Stockwell
ABSTRACT
Precision medicine in oncology requires not only identification of cancer-associated mutations but also effective drugs for each cancer genotype, which is still a largely unsolved problem. One approach for the latter challenge has been large-scale testing of small molecules in genetically characterized cell lines. We hypothesized that compounds with high cell-line-selective lethality exhibited consistent results across such pharmacogenomic studies. We analyzed the compound sensitivity data of 6,259 lethal compounds from the NCI-60 project. A total of 2,565 cell-line-selective lethal compounds were identified and grouped into 18 clusters based on their median growth inhibitory GI50 profiles across the 60 cell lines, which were shown to represent distinct mechanisms of action. Further transcriptome analysis revealed a biomarker, NADPH abundance, for predicting sensitivity to ferroptosis-inducing compounds, which we experimentally validated. In summary, incorporating cell-line-selectivity filters improves the predictive power of pharmacogenomic analyses and enables discovery of biomarkers that predict the sensitivity of cells to specific cell death inducers.
Sigma-Aldrich
ML162, ≥98% (HPLC)