MilliporeSigma
Lower urinary tract infections in women.
British journal of nursing (Mark Allen Publishing) (2017-05-12)
Cathy Davis, Angie Rantell
ABSTRACT
In her lifetime, a woman is highly likely to develop at least one lower urinary tract infection. Early detection and treatment are key. Being aware of predisposing factors for infection and understanding appropriate diagnosis and treatment regimens will help nurses in both primary and acute care manage these patients correctly. This will not only benefit patients but will also help prevent incorrect antimicrobial management and avoid unplanned admissions. This aim of this article is to provide nurses with the information they need to best advise both colleagues and patients on how to manage lower urinary tract infections in women.
Sigma-Aldrich
Methenamine hippurate, ≥95% (HPLC)