MilliporeSigma
  • Triazolopyrimidines Are Microtubule-Stabilizing Agents that Bind the Vinca Inhibitor Site of Tubulin.
Triazolopyrimidines Are Microtubule-Stabilizing Agents that Bind the Vinca Inhibitor Site of Tubulin.
Cell chemical biology (2017-06-06)
Gonzalo Sáez-Calvo, Ashwani Sharma, Francisco de Asís Balaguer, Isabel Barasoain, Javier Rodríguez-Salarichs, Natacha Olieric, Hugo Muñoz-Hernández, Manuel Álvaro Berbís, Sebastian Wendeborn, Miguel Angel Peñalva, Ruth Matesanz, Ángeles Canales, Andrea Enrico Prota, Jesús Jímenez-Barbero, José Manuel Andreu, Clemens Lamberth, Michel Olivier Steinmetz, José Fernando Díaz
ABSTRACT
Microtubule-targeting agents (MTAs) are some of the clinically most successful anti-cancer drugs. Unfortunately, instances of multidrug resistances to MTA have been reported, which highlights the need for developing MTAs with different mechanistic properties. One less explored class of MTAs are [1,2,4]triazolo[1,5-a]pyrimidines (TPs). These cytotoxic compounds are microtubule-stabilizing agents that inexplicably bind to vinblastine binding site on tubulin, which is typically targeted by microtubule-destabilizing agents. Here we used cellular, biochemical, and structural biology approaches to address this apparent discrepancy. Our results establish TPs as vinca-site microtubule-stabilizing agents that promote longitudinal tubulin contacts in microtubules, in contrast to classical microtubule-stabilizing agents that primarily promote lateral contacts. Additionally we observe that TPs studied here are not affected by p-glycoprotein overexpression, and suggest that TPs are promising ligands against multidrug-resistant cancer cells.
Sigma-Aldrich
Cevipabulin fumarate, ≥98% (HPLC)