MilliporeSigma
  • Mechanism of action of the antineoplastic drug lonidamine: 31P and 13C nuclear magnetic resonance studies.
Mechanism of action of the antineoplastic drug lonidamine: 31P and 13C nuclear magnetic resonance studies.
Cancer research (1995-07-01)
H Ben-Horin, M Tassini, A Vivi, G Navon, O Kaplan
ABSTRACT
The mechanism of action of the antineoplastic drug lonidamine (LND) on MCF-7 human breast cancer cells was studied with the use of 31P and 13C nuclear magnetic resonance (NMR) spectroscopy. The cells were embedded in alginate microcapsules, perfused with growth media and LND at physiological conditions in the NMR tube, and continuously monitored in vivo for the effects of LND. 31P NMR demonstrated intracellular acidification after LND perfusion concomitant with ATP depletion and changes in phospholipid metabolites. 13C NMR showed marked LND-induced accumulation of lactate, and spectra of the perfusate disclosed that LND inhibited lactate transport. Kinetic 13C NMR also furnished information on LND effects on glucose metabolism; LND decreased initial glucose uptake and lactate formation, although the final intracellular glucose levels were higher compared with those in controls. Combined administration of LND and the metabolic inhibitor 2-deoxyglucose yielded additive but not synergistic cytotoxicity and enabled assessment of hexokinase activity. Overall, the results indicate that the major metabolic changes induced by LND are inhibition of lactate transport and its accumulation, which lead to intracellular acidification.
Sigma-Aldrich
Lonidamine, mitochondrial hexokinase inhibitor