MilliporeSigma
  • Murine alpha-N-acetylgalactosaminidase: isolation and expression of a full-length cDNA and genomic organization: further evidence of an alpha-galactosidase gene family.
Murine alpha-N-acetylgalactosaminidase: isolation and expression of a full-length cDNA and genomic organization: further evidence of an alpha-galactosidase gene family.
Molecular genetics and metabolism (1998-10-27)
A M Wang, Y A Ioannou, K M Zeidner, R J Desnick
ABSTRACT
Recent characterization of the human sequences encoding two lysosomal hydrolases, alpha-galactosidase A (alpha-Gal A) and alpha-N-acetylgalactosaminidase (alpha-GalNAc) revealed that these two enzymes with distinct enzymatic activities shared about 50% overall amino acid identity and that their genomic sequences had a conserved common gene structure. These findings suggested that these genes, which are located on different chromosomes, arose by duplication and divergence from a common ancestral gene. To further compare this alpha-galactosidase gene family, the murine alpha-GalNAc cDNA and genomic sequences were isolated and characterized. The full-length cDNA contained an open-reading frame of 1245 bp encoding a 415 amino acid polypeptide and had 5' and 3' untranslated regions of 94 and 333 bp, respectively. The coding region had 81% nucleotide and 81.9% amino acid identities with those of the corresponding human alpha-GalNAc sequence. Northern analysis revealed a single transcript of approximately 1.9 kb. The functional integrity of the cDNA was demonstrated by transient expression in COS-1 cells. The murine alpha-GalNAc genomic sequence spanned approximately 9 kb and was identical in structure with the human alpha-GalNAc gene with eight introns interrupting the coding sequence at identical positions. In addition, the deduced amino acid sequence of the murine alpha-GalNAc gene was highly homologous with alpha-GalNAc and alpha-Gal A genes from other species providing further support for a common evolutionary ancestor of the alpha-galactosidase gene family. The availability of the murine gene will permit additional evolutionary comparisons, structure/function analyses, and the generation of mice with alpha-GalNAc deficiency by gene targeting.
Sigma-Aldrich
Endo-α-N-acetylgalactosaminidase, Streptococcus pneumoniae, Recombinant, E. coli, Endo-α-N-acetylgalactosaminidase, Streptococcus pneumoniae, Recombinant, E. coli, CAS 59793-96-3, catalyzes the hydrolysis of the unsubstituted Galβ1,3GalNAc core disaccharide attached to Ser or Thr.