MilliporeSigma
  • Okadaic acid enhances human T cell activation and phosphorylation of an internal substrate induced by phorbol myristate acetate.
Okadaic acid enhances human T cell activation and phosphorylation of an internal substrate induced by phorbol myristate acetate.
Immunopharmacology (1992-07-01)
Y Tada, S Yoshizawa, K Nagasawa, I Furugo, T Tsuru, T Mayumi, H Tsukamoto, Y Niho
ABSTRACT
Okadaic acid is a potent tumor promoter and an inhibitor of serine/threonine-specific protein phosphatases. We studied the effect of okadaic acid in human T cell activation and phosphorylation of internal substrates. Okadaic acid at up to 4 nM enhanced phorbol myristate acetate (PMA)-induced proliferation and CD25 (IL-2 receptor, p55) expression, although it showed no activation by itself. Okadaic acid induced hyperphosphorylation of a 60 kDa protein in T cells as well as non-T cells, as reported in fibroblasts and keratinocytes. Preincubation with 4 nM okadaic acid enhanced PMA induced phosphorylation of the 80 kDa protein, an internal substrate of protein kinase C in T cells. These results suggest that okadaic acid inhibited dephosphorylation of protein kinase C specific substrates, and as a result, enhanced T cell activation mediated by protein kinase C pathway.
Sigma-Aldrich
Okadaic Acid, Sodium Salt, Water-soluble analog of Okadaic Acid. Inhibits protein phosphatases 1 and 2A.