MilliporeSigma
  • Inhibition of inducible nitric oxide synthase gene expression by indomethacin or ibuprofen in beta-amyloid protein-stimulated J774 cells.
Inhibition of inducible nitric oxide synthase gene expression by indomethacin or ibuprofen in beta-amyloid protein-stimulated J774 cells.
European journal of pharmacology (2000-11-18)
O Ogawa, H Umegaki, D Sumi, T Hayashi, A Nakamura, N K Thakur, J Yoshimura, H Endo, A Iguchi
ABSTRACT
Recent studies show that a mononuclear phagocyte lineage, including microglia, plays a possible role in the pathogenesis of Alzheimer's disease through nitric oxide (NO)-mediated neurotoxicity. Epidemiological studies show that nonsteroidal anti-inflammatory drugs (NSAIDs) have a protective effect against Alzheimer's disease. Based on these observations, it has been hypothesized that an anti-Alzheimer's disease effect of NSAIDs could result from the inhibition of NO synthesis. We report here that indomethacin or ibuprofen dose-dependently reduce beta-amyloid protein and interferon-gamma-induced NO production, accompanied by an inhibition of inducible nitric oxide synthase mRNA expression in J774 cells, a murine macrophage cell line. Aspirin, however, does not produce such an effect, suggesting that the cyclooxygenases pathway is not involved in the inhibitory effects of NSAIDs on beta-amyloid protein and interferon-gamma-induced NO production in J774 cells.
Sigma-Aldrich
(±)-Ibuprofen, A nonsteroidal anti-inflammatory drug (NSAID) that acts as a reversible and competitive inhibitor of cyclooxygenase 1 (COX-1) (IC₅₀ = 4.85 µM).