MilliporeSigma
  • BcXyl, a β-xylosidase Isolated from Brunfelsia Calycina Flowers with Anthocyanin-β-glycosidase Activity.
BcXyl, a β-xylosidase Isolated from Brunfelsia Calycina Flowers with Anthocyanin-β-glycosidase Activity.
International journal of molecular sciences (2019-03-25)
Boyu Dong, Honghui Luo, Bin Liu, Wenjun Li, Shaojian Ou, Yongyi Wu, Xuelian Zhang, Xuequn Pang, Zhaoqi Zhang
ABSTRACT
Brunfelsia calycina flowers lose anthocyanins rapidly and are therefore well suited for the study of anthocyanin degradation mechanisms, which are unclear in planta. Here, we isolated an anthocyanin-β-glycosidase from B. calycina petals. The MS/MS (Mass Spectrometry) peptide sequencing showed that the enzyme (72 kDa) was a β-xylosidase (BcXyl). The enzyme showed high activity to p-Nitrophenyl-β-d-galactopyranoside (pNPGa) and p-Nitrophenyl-β-d-xylopyranoside (pNPX), while no activity to p-Nitrophenyl-β-d-glucopyranoside (pNPG) or p-Nitrophenyl-β-D-mannopyranoside (pNPM) was seen. The optimum temperature of BcXyl was 40 °C and the optimum pH was 5.0. The enzyme was strongly inhibited by 1 mM D-gluconate and Ag⁺. HPLC (High Performance Liquid Chromatography) analysis showed that BcXyl catalyzed the degradation of an anthocyanin component of B. calycina, and the release of xylose and galactose due to hydrolysis of glycosidic bonds by BcXyl was detected by GC (Gas Chromatography) /MS. A full-length mRNA sequence (2358 bp) of BcXyl (NCBI No. MK411219) was obtained and the deduced protein sequence shared conserved domains with two anthocyanin-β-glycosidases (Bgln and BadGluc, characterized in fungi). BcXyl, Bgln and BadGluc belong to AB subfamily of Glycoside hydrolase family 3. Similar to BcPrx01, an anthocyanin-degradation-related Peroxidase (POD), BcXyl was dramatically activated at the stage at which the rapid anthocyanin degradation occurred. Taken together, we suggest that BcXyl may be the first anthocyanin-β-glycosidase identified in higher plants.