MilliporeSigma
  • Rhus and Safflower Extracts as Potential Novel Food Antioxidant, Anticancer, and Antimicrobial Agents Using Nanotechnology.
Rhus and Safflower Extracts as Potential Novel Food Antioxidant, Anticancer, and Antimicrobial Agents Using Nanotechnology.
Foods (Basel, Switzerland) (2019-04-26)
Faten Y Ibrahim, Ayman Y El-Khateeb, Azza H Mohamed
ABSTRACT
Green synthesis of metal nanoparticles using plant extracts offers a safe and attractive alternate to the chemical methods. The present work aims at preparing metal nanoparticles of rhus (Rhus coriaria L.) and safflower (Carthamus tinctorius L.) extracts using Fe2+, Cu2+, Zn2+, and Ag+ ions. The water extracts were prepared, and the total polyphenols and flavonoids contents were determined. The safflower extract contained the highest number of total polyphenols and total flavonoids (87.20 mg GAE/g and 36.32 mg QE/g), respectively. The synthesized nanoparticles were characterized using UV-Visible (UV-Vis) spectroscopy and Transmission Electron Microscope (TEM). The studied extracts and their nanoparticles were evaluated as an antioxidant, antimicrobial, and anticancer agents. The plant extracts and their nanoparticles showed significant antioxidant activity using (3-ethylbenzothiazoline-6-sulfonic acid (ABTS•+) and 2, 2-diphenyl-1-picrylhydrazyl (DPPH) assays. Safflower silver nanoparticles (AgNPs) were the most powerful antimicrobial agent compared to the other nanoparticles. The Sulforhodamine B (SRB) cytotoxic activity was evaluated against three cancer cell lines. The results revealed that CuNP safflower nanoparticles displayed the highest activity as anticancer agent with values (98.94% with T47D, 97.68% with HEPG2, and 89.33% against Caco-2). The data revealed that rhus and safflower extracts and their nanoparticles possess high potential activity as antimicrobial, antioxidant, and anticancer agents.