MilliporeSigma
  • Nesprin-2G knockout fibroblasts exhibit reduced migration, changes in focal adhesion composition, and reduced ability to generate traction forces.
Nesprin-2G knockout fibroblasts exhibit reduced migration, changes in focal adhesion composition, and reduced ability to generate traction forces.
Cytoskeleton (Hoboken, N.J.) (2019-01-23)
Alexandra Woychek, Jonathan C R Jones
ABSTRACT
The nuclear envelope protein nesprin-2G is a component of the linker of nucleoskeleton and cytoskeleton (LINC) complex and is responsible for mechanical and signaling crosstalk between the nucleus and cytoskeleton. A prior study has demonstrated that nesprin-2G knockout mice show delayed wound healing. The goal was to elucidate the mechanism underlying the delayed wound closure in this mouse model. Primary fibroblasts from wild-type and knockout neonatal mice were isolated. Knockout cells exhibited decreased focal adhesion (FA) size, number, and intensity. Consistent with this result, FA protein expression levels were decreased in knockout cells. Additionally, knockout fibroblasts displayed an abnormal actin cytoskeleton, as evidenced by loss of TAN line formation and both cytoplasmic and peri-nuclear actin staining. Using collective and single cell motility assays, it was found that knockout cells exhibited a reduction in both speed and directed migration. Traction force microscopy revealed that knockout fibroblasts generated fewer traction forces compared with WT fibroblasts. In summary, the data indicated that changes in actin organization and defects in FAs result in a reduced ability of knockout fibroblasts to generate traction forces needed for efficient motility.
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
Anti-Zyxin antibody, Mouse monoclonal, clone ZOL301, purified from hybridoma cell culture
Sigma-Aldrich
Anti-GAPDH Antibody, from chicken, purified by affinity chromatography