MilliporeSigma
  • Inhibition of Dipeptidyl Peptidase-4 Accelerates Epithelial-Mesenchymal Transition and Breast Cancer Metastasis via the CXCL12/CXCR4/mTOR Axis.
Inhibition of Dipeptidyl Peptidase-4 Accelerates Epithelial-Mesenchymal Transition and Breast Cancer Metastasis via the CXCL12/CXCR4/mTOR Axis.
Cancer research (2018-12-26)
Fan Yang, Yuta Takagaki, Yasuo Yoshitomi, Takayuki Ikeda, Jinpeng Li, Munehiro Kitada, Asako Kumagai, Emi Kawakita, Sen Shi, Keizo Kanasaki, Daisuke Koya
ABSTRACT
Dipeptidyl peptidase (DPP)-4 is a multifunctional glycoprotein involved in various biological and pathologic processes. DPP-4 has been widely recognized as a therapeutic target for type 2 diabetes mellitus but is also implicated in the development of human malignancies. Here, we show that inhibition of DPP-4 accelerates breast cancer metastasis via induction of CXCL12/CXCR4, which activates mTOR to promote epithelial-mesenchymal transition (EMT). In cultured cells, DPP-4 knockdown induced EMT and cell migration. Treatment with the DPP-4 inhibitor KR62436 (KR) promoted primary tumor growth and lung metastasis in a 4T1 tumor allograft mouse model; DPP-4 knockdown in 4T1 cells displayed similar phenotypes in vivo and in vitro. KR treatment enhanced the levels of CXCL12/CXCR4 and phosphorylated mTOR, which were associated with the induction of EMT in metastatic cancer cells. KR-induced EMT in cancer cells was inhibited by treatment with the CXCR4 inhibitor AMD3100 or the mTOR inhibitor rapamycin, and AMD3100 suppressed KR-induced metastasis in vivo. Our findings suggest that DPP-4 plays a significant role in cancer biology and that inhibition of DPP-4 promotes cancer metastasis via induction of the CXCL12/CXCR4/mTOR/EMT axis. SIGNIFICANCE: These findings reveal that inhibition of DPP-4 increases the metastatic potential of breast cancer. This is especially important given the potential use of DPP-4 inhibition as a therapeutic strategy for type 2 diabetes.
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Anti-TGF β Receptor I antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-DPP4/CD26 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution