MilliporeSigma
  • The Prevention of Hepatectomy-Induced Adhesions by Bilayer Sponge Composed of Ultrapure Alginate.
The Prevention of Hepatectomy-Induced Adhesions by Bilayer Sponge Composed of Ultrapure Alginate.
The Journal of surgical research (2019-05-28)
Seiichi Ohta, Takeo Toda, Fuyuki Inagaki, Kiyohiko Omichi, Atsushi Shimizu, Norihiro Kokudo, Kiyoshi Hasegawa, Taichi Ito
ABSTRACT
Adhesion formation is a critical issue in surgery, particularly in hepatectomy. The present study aimed to develop a bilayer adhesion barrier comprising alginate (Alg) of different molecular weight (Mw). It was expected that a slowly dissolving layer remains on the cut surface, functioning as a physical barrier, whereas a rapidly dissolving layer widely distributes in the peritoneal cavity to prevent de novo adhesions. Bilayer Alg sponges were fabricated using low Mw Alg for the upper layer and high Mw Alg for the bottom layer. The dissolution behavior of each layer was evaluated in vitro in peritoneum-like environments. We constructed a Pean crush hepatectomy-induced adhesion model in rats. The effects of the bilayer sponge on cut surface and de novo adhesions were separately evaluated in terms of their extent and grade. The Alg sponge layer with low Mw dissolved faster than that with high Mw in vitro. One week after the hepatectomy, although no significant decrease in adhesion extent on the cut surface was observed in rats that received Seprafilm and Interceed, treatment with Alg bilayer sponge significantly decreased the adhesion extent to 38% of that without treatment. Moreover, a significant decrease in de novo adhesion extent was observed in the Alg bilayer sponge compared with the Interceed group. The Alg bilayer sponge was effective for preventing both cut surface and de novo adhesions in the rat Pean crush hepatectomy model. The simple yet functional design of the Alg bilayer sponge can facilitate its use in future clinical practice.