- Heterocycle Effects on the Liquid Crystallinity of Terthiophene Analogues.
Heterocycle Effects on the Liquid Crystallinity of Terthiophene Analogues.
Liquid crystalline self-assembly offers the potential to create highly ordered, uniformly aligned, and defect-free thin-film organic semiconductors. Analogues of one of the more promising classes of liquid crystal semiconductors, 5,5"-dialkyl-α-terthiophenes, were prepared in order to investigate the effects of replacing the central thiophene with either an oxadiazole or a thiadiazole ring. The phase behaviour was examined by differential scanning calorimetry, polarized optical microscopy, and variable temperature x-ray diffraction. While the oxadiazole derivative was not liquid crystalline, thiadiazole derivatives formed smectic C and soft crystal lamellar phases, and maintained lamellar order down to room temperature. Variation of the terminal alkyl chains also influenced the observed phase sequence. Single crystal structures revealed the face-to-face orientation of molecules within the layers in the solid-state, a packing motif that is rationalized based on the shape and dipole of the thiadiazole ring, as corroborated by density functional theory (DFT) calculations. The solution opto-electronic properties of the systems were characterized by absorption and emission spectroscopy, cyclic voltammetry, and time-dependent density functional theory (TD-DFT).