MilliporeSigma
  • Vapor-printed polymer electrodes for long-term, on-demand health monitoring.
Vapor-printed polymer electrodes for long-term, on-demand health monitoring.
Science advances (2019-03-23)
Jae Joon Kim, Linden K Allison, Trisha L Andrew
ABSTRACT
We vapor print conformal conjugated polymer electrodes directly onto living plants and use these electrodes to probe the health of actively growing specimens using bioimpedance spectroscopy. Vapor-printed polymer electrodes, unlike their adhesive thin-film counterparts, do not delaminate from microtextured living surfaces as the organism matures and do not observably attenuate the natural growth pattern and self-sustenance of the plants investigated here. On-demand, noninvasive bioimpedance spectroscopy performed with long-lasting vapor-printed polymer electrodes can reliably detect deep tissue damage caused by dehydration and ultraviolet A exposure throughout the life cycle of a plant.