MilliporeSigma
  • Evaluation of sulfate metabolites as markers of intramuscular testosterone administration in Caucasian and Asian populations.
Evaluation of sulfate metabolites as markers of intramuscular testosterone administration in Caucasian and Asian populations.
Drug testing and analysis (2019-04-02)
Argitxu Esquivel, Élida Alechaga, Nuria Monfort, Sheng Yang, Yanyi Xing, Wu Moutian, Rosa Ventura
ABSTRACT
The introduction of alternative markers to the steroid profile can be an effective approach to improving the screening capabilities for the detection of testosterone (T) misuse. In this work, endogenous steroid sulfates were evaluated as potential markers to detect intramuscular (IM) T administration. Fourteen sulfate metabolites were quantified using mixed-mode solid-phase extraction and analysis by liquid chromatography-tandem mass spectrometry (LC-MS/MS). Urine samples after a single IM injection (100 mg) of T cypionate to six Caucasian and six Asian healthy male volunteers were analyzed. Principal component analysis (PCA) was used to characterize the sample cohort and to obtain the most useful markers for discrimination between pre- and post-administration samples. For Caucasian volunteers, a separation between pre- and post-administration samples was observed in PCA, whereas for Asian volunteers no separation was obtained. Seventeen ratios between sulfate metabolites were selected and further considered. Detection times (DTs) of each marker were evaluated using individual thresholds for each volunteer. The best results were obtained using ratios involving T and epitestosterone (E) sulfates in the denominator. The best marker was the ratio androsterone sulfate/testosterone sulfate (A-S/T-S) which prolonged the DT 1.2-2.1 times in respect to those obtained using T/E ratio in all Caucasian volunteers and 1.3-1.5 times in two Asian volunteers. Other ratios between A-S or etiocholanolone sulfate and E-S, and sulfates of etiocholanolone, dehydroandrosterone or epiandrosterone, and T-S were also found adequate. These ratios improve the DT after IM T administration and their incorporation to complement the current steroid profile is recommended.