MilliporeSigma
  • Novel biotechnological glucosylation of high-impact aroma chemicals, 3(2H)- and 2(5H)-furanones.
Novel biotechnological glucosylation of high-impact aroma chemicals, 3(2H)- and 2(5H)-furanones.
Scientific reports (2019-07-31)
Isabelle Effenberger, Thomas Hoffmann, Rafal Jonczyk, Wilfried Schwab
ABSTRACT
Glucosyltransferases are versatile biocatalysts to chemically modify small molecules and thus enhance their water solubility and structural stability. Although the genomes of all organisms harbor a multitude of glucosyltransferase genes, their functional characterization is hampered by the lack of high-throughput in-vivo systems to rapidly test the versatility of the encoded proteins. We have developed and applied a high-throughput whole cell biotransformation system to screen a plant glucosyltransferase library. As proof of principle, we identified 25, 24, 15, and 18 biocatalysts transferring D-glucose to sotolone, maple furanone, furaneol and homofuraneol, four highly appreciated flavor compounds, respectively. Although these 3(2H)- and 2(5H)-furanones have extremely low odor thresholds their glucosides were odorless. Upscaling of the biotechnological process yielded titers of 5.3 and 7.2 g/L for the new to nature β-D-glucopyranosides of sotolone and maple furanone, respectively. Consequently, plant glucosyltransferase show stunning catalytic activities, which enable the economical production of novel and unexplored chemicals with exciting new functionalities by whole-cell biotransformation.