MilliporeSigma
  • Effects of PPARs/20-HETE on the renal impairment under diabetic conditions.
Effects of PPARs/20-HETE on the renal impairment under diabetic conditions.
Experimental cell research (2019-06-05)
Shumei Ding, Jiajun Huang, Hongmei Qiu, Rongchun Chen, Jie Zhang, Bo Huang, Oumei Cheng, Qingsong Jiang
ABSTRACT
Diabetic nephropathy (DN) is one of the most severe complications of diabetes mellitus. The pathomolecular events behind DN remain uncertain. Peroxisome proliferator-activated receptors (PPARs) play essential functions in the development of DN. Meanwhile, 20-hydroxyeicosatetraenoic acid (20-HETE) also plays central roles in the regulation of renal function. However, the relationship between PPARs and 20-HETE is rarely studied in DN. It was revealed in our study that both PPARs expression and CYP4A-20-HETE level were decreased under DN conditions in vivo and in vitro. Supplementation with bezafibrate, a PPAR pan-agonist, improved the damage of kidney in DN mice and in high glucose-induced NRK-52E cells, following the up-regulation of PPARs and the increase of CYP4A-20-HETE. PPARα antagonist (MK886), PPARβ antagonist (GSK0660), and PPARγ antagonist (GW9662) reversed the protection of bezafibrate in NRK-52E, and abrogated the up-regulation of CYP4A-20-HETE produced by bezafibrate. Noteworthily, 20-HETE synthetase inhibitor, HET0016, also blocked the bezafibrate-mediated improvement of NRK-52E, and abolished the up-regulation of PPARs expression. Collectively, our data suggest that the concurrent down-regulation and interaction of PPARs and 20-HETE play crucial roles in the pathogenesis process of DN, and we provide a novel evidence that PPARs/20-HETE signaling may be served as a therapeutic target for DN patients.