MilliporeSigma
  • G9a/GLP Complex Acts as a Bidirectional Switch to Regulate Metabotropic Glutamate Receptor-Dependent Plasticity in Hippocampal CA1 Pyramidal Neurons.
G9a/GLP Complex Acts as a Bidirectional Switch to Regulate Metabotropic Glutamate Receptor-Dependent Plasticity in Hippocampal CA1 Pyramidal Neurons.
Cerebral cortex (New York, N.Y. : 1991) (2018-07-10)
Mahima Sharma, Sreedharan Sajikumar
ABSTRACT
Metabotropic glutamate receptor-dependent long-term depression (mGluR-LTD) is conventionally considered to be solely dependent on local protein synthesis. Given the impact of epigenetics on memory, the intriguing question is whether epigenetic regulation influences mGluR-LTD as well. G9a/GLP histone lysine methyltransferase complex is crucial for brain development and goal-directed learning as well as for drug-addiction. In this study, we analyzed whether the epigenetic regulation by G9a/GLP complex affects mGluR-LTD in CA1 hippocampal pyramidal neurons of 5-7 weeks old male Wistar rats. In hippocampal slices with intact CA1 dendritic regions, inhibition of G9a/GLP activity abolished mGluR-LTD. The inhibition of this complex upregulated the expression of plasticity proteins like PKMζ, which mediated the prevention of mGluR-LTD expression by regulating the NSF-GluA2-mediated trafficking of AMPA receptors towards the postsynaptic site. G9a/GLP inhibition during the induction of mGluR-LTD also downregulated the protein levels of phosphorylated-GluA2 and Arc. Interestingly, G9a/GLP inhibition could not impede the mGluR-LTD when the cell-body was severed. Our study highlights the role of G9a/GLP complex in intact neuronal network as a bidirectional switch; when turned on, it facilitates the expression of mGluR-LTD, and when turned off, it promotes the expression of long-term potentiation.