MilliporeSigma
  • Presence of pharmaceuticals in fish collected from urban rivers in the U.S. EPA 2008-2009 National Rivers and Streams Assessment.
Presence of pharmaceuticals in fish collected from urban rivers in the U.S. EPA 2008-2009 National Rivers and Streams Assessment.
The Science of the total environment (2018-04-11)
Belinda Huerta, Sara Rodriguez-Mozaz, Jim Lazorchak, Damia Barcelo, Angela Batt, John Wathen, Leanne Stahl
ABSTRACT
Fish are good indicators of aquatic environment pollution because of their capability to uptake pollutants contained in water. Therefore, accumulation of pharmaceutical compounds in freshwater and marine fish and other aquatic organisms has been studied extensively in the last decade. In this context, the present study investigates the occurrence of pharmaceutical compounds in wild fish from 25 polluted river sites in the USA, downstream from wastewater treatment plants (WWTPs). Sample sites constitute a subset of urban rivers investigated in the U.S. EPA's 2008-2009 National Rivers and Streams Assessment. Thirteen pharmaceuticals (out of the twenty compounds analyzed) were quantified in fish fillets at concentrations commonly below 10ngg-1, in accordance with the findings from previous studies in the USA and Europe. The psychoactive drugs venlafaxine, carbamazepine and its metabolite 2-hydroxy carbamazepine were the most prevalent compounds (58%, 27% and 42%, respectively). This group of drugs is highly prescribed and rather resistant to degradation during conventional treatment in WWTPs as well as in natural aquatic environments. Salbutamol, a drug used to treat asthma, and the diuretic hydrochlorothiazide were also frequently detected (in >20% of the samples). Occurrence of six pharmaceutical families due to chronic exposure at environmental concentrations in water was detected in eight fish species.