MilliporeSigma
  • A KLF6-driven transcriptional network links lipid homeostasis and tumour growth in renal carcinoma.
A KLF6-driven transcriptional network links lipid homeostasis and tumour growth in renal carcinoma.
Nature communications (2019-03-13)
Saiful E Syafruddin, Paulo Rodrigues, Erika Vojtasova, Saroor A Patel, M Nazhif Zaini, Johanna Burge, Anne Y Warren, Grant D Stewart, Tim Eisen, Dóra Bihary, Shamith A Samarajiwa, Sakari Vanharanta
ABSTRACT
Transcriptional networks are critical for the establishment of tissue-specific cellular states in health and disease, including cancer. Yet, the transcriptional circuits that control carcinogenesis remain poorly understood. Here we report that Kruppel like factor 6 (KLF6), a transcription factor of the zinc finger family, regulates lipid homeostasis in clear cell renal cell carcinoma (ccRCC). We show that KLF6 supports the expression of lipid metabolism genes and promotes the expression of PDGFB, which activates mTOR signalling and the downstream lipid metabolism regulators SREBF1 and SREBF2. KLF6 expression is driven by a robust super enhancer that integrates signals from multiple pathways, including the ccRCC-initiating VHL-HIF2A pathway. These results suggest an underlying mechanism for high mTOR activity in ccRCC cells. More generally, the link between super enhancer-driven transcriptional networks and essential metabolic pathways may provide clues to the mechanisms that maintain the stability of cell identity-defining transcriptional programmes in cancer.
Sigma-Aldrich
Fatostatin hydrobromide, ≥98% (HPLC), powder
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Simvastatin, ≥97% (HPLC), solid
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture