MilliporeSigma
  • Adaptation and tolerance mechanisms developed by mycorrhizal Bipinnula fimbriata plantlets (Orchidaceae) in a heavy metal-polluted ecosystem.
Adaptation and tolerance mechanisms developed by mycorrhizal Bipinnula fimbriata plantlets (Orchidaceae) in a heavy metal-polluted ecosystem.
Mycorrhiza (2018-08-11)
Héctor Herrera, Rafael Valadares, Guilherme Oliveira, Alejandra Fuentes, Leonardo Almonacid, Sidney Vasconcelos do Nascimento, Yoav Bashan, Cesar Arriagada
ABSTRACT
The adaptation and performance of orchid mycorrhizae in heavy metal-polluted soils have been poorly explored. In the present study, proteomic and metabolic approaches were used to detect physiological changes in orchid roots established in a heavy metal-polluted soil and to ascertain whether mycorrhizal fungi affect the metabolic responses of roots. Young Bipinnula fimbriata plantlets were established in control and heavy metal-polluted soils in a greenhouse. After 14 months, exudation of root organic acids, phenolics, percentage of mycorrhization, mineral content, and differential protein accumulation were measured. More root biomass, higher root colonization, and higher exudation rates of citrate, succinate, and malate were detected in roots growing in heavy metal-polluted soils. Higher accumulation of phosphorus and heavy metals was found inside mycorrhizal roots under metal stress. Under non-contaminated conditions, non-mycorrhizal root segments showed enhanced accumulation of proteins related to carbon metabolism and stress, whereas mycorrhizal root segments stimulated protein synthesis related to pathogen control, cytoskeleton modification, and sucrose metabolism. Under heavy metal stress, the proteome profile of non-mycorrhizal root segments indicates a lower induction of defense mechanisms, which, together with the stimulation of enzymes related to carotenoid biosynthesis and cell wall organization, may positively influence mycorrhizal fungi colonization. The results point to different metabolic strategies in mycorrhizal and non-mycorrhizal root segments that are exposed to heavy metal stress. The results indicate that root colonization by mycorrhizal fungi is stimulated to alleviate the negative effects of heavy metals in the orchids.