MilliporeSigma
  • Synthesis, Characterisation and In Vitro Permeation, Dissolution and Cytotoxic Evaluation of Ruthenium(II)-Liganded Sulpiride and Amino Alcohol.
Synthesis, Characterisation and In Vitro Permeation, Dissolution and Cytotoxic Evaluation of Ruthenium(II)-Liganded Sulpiride and Amino Alcohol.
Scientific reports (2019-03-13)
Gretta C M'bitsi-Ibouily, Thashree Marimuthu, Pradeep Kumar, Yahya E Choonara, Lisa C du Toit, Priyamvada Pradeep, Girish Modi, Viness Pillay
ABSTRACT
Sulpiride (SPR) is a selective antagonist of central dopamine receptors but has limited clinical use due to its poor pharmacokinetics. The aim of this study was to investigate how metal ligation to SPR may improve its solubility, intestinal permeability and prolong its half-life. The synthesis and characterisation of ternary metal complexes [Ru(p -cymene)(L)(SPR)]PF6 (L1 = (R)-(+)-2-amino-3-phenyl-1-propanol, L2 = ethanolamine, L3 = (S)-(+)-2-amino-1-propanol, L4 = 3-amino-1-propanol, L5 = (S)-(+)-2-pyrrolidinemethanol) are described in this work. The stability constant of the [Ru(p -cymene)(SPR)] complex was determined using Job's method. The obtained value revealed higher stability of the metal complex in the physiological pH than in an acidic environment such as the stomach. The ternary metal complexes were characterised by elemental analysis, Fourier transform infrared spectroscopy (FT-IR), 1H and 13C nuclear magnetic resonance (NMR), differential scanning calorimetry (DSC), thermal analyses, Ultraviolet-Visible (UV-Vis). Solubility studies showed higher aqueous solubility for complexed SPR than the free drug. Dissolution profiles of SPR from the metal complexes exhibited slower dissolution rate of the drug. Permeation studies through the pig's intestine revealed enhanced membrane permeation of the complexed drug. In vitro methyl thiazolyl tetrazolium (MTT) assay showed no noticeable toxic effects of the ternary metal complexes on Caco-2 cell line.