MilliporeSigma
  • Strong Surface-Bound Sulfur in Carbon Nanotube Bridged Hierarchical Mo2 C-Based MXene Nanosheets for Lithium-Sulfur Batteries.
Strong Surface-Bound Sulfur in Carbon Nanotube Bridged Hierarchical Mo2 C-Based MXene Nanosheets for Lithium-Sulfur Batteries.
Small (Weinheim an der Bergstrasse, Germany) (2018-12-18)
Li-Ping Lv, Chao-Fei Guo, Weiwei Sun, Yong Wang
ABSTRACT
In this work, hydroxyl-functionalized Mo2 C-based MXene nanosheets are synthesized by facilely removing the Sn layer of Mo2 SnC. The hydroxyl-functionalized surface of Mo2 C suppresses the shuttle effect of lithium polysulfides (LiPSs) through strong interaction between Mo atoms on the MXenes surface and LiPSs. Carbon nanotubes (CNTs) are further introduced into Mo2 C phase to enlarge the specific surface area of the composite, improve its electronic conductivity, and alleviate the volume change during discharging/charging. The strong surface-bound sulfur in the hierarchical Mo2 C-CNTs host can lead to a superior electrochemical performance in lithium-sulfur batteries. A large reversible capacity of ≈925 mAh g-1 is observed after 250 cycles at a current density of 0.1 C (1 C = 1675 mAh g-1 ) with good rate capability. Notably, the electrodes with high loading amounts of sulfur can also deliver good electrochemical performances, i.e., initial reversible capacities of ≈1314 mAh g-1 (2.4 mAh cm-2 ), ≈1068 mAh g-1 (3.7 mAh cm-2 ), and ≈959 mAh g-1 (5.3 mAh cm-2 ) at various areal loading amounts of sulfur (1.8, 3.5, and 5.6 mg cm-2 ) are also observed, respectively.