MilliporeSigma
  • Swine model of in-stent stenosis in the iliac artery evaluating the serial time course.
Swine model of in-stent stenosis in the iliac artery evaluating the serial time course.
Experimental animals (2018-08-03)
Osamu Ishikawa, Minoru Tanaka, Kenjiro Konno, Terumitsu Hasebe, Ayumi Horikawa, Akira Iijima, Nobuhito Saito, Koki Takahashi
ABSTRACT
The aim of this study was to propose a new animal model evaluating the serial time course of in-stent stenosis by repeated carotid artery catheterization in the same animal. 16 bare-metal stents were implanted in the normal external and internal iliac artery of 8 miniature pigs. Repeated measurements were performed in the same animal every 2 weeks for 12 weeks through carotid artery catheterization. The time course and peak neointimal proliferation were evaluated by intravascular ultrasound. Health of all animals was assessed by clinical and hematological examinations. As a result, 7 times of carotid artery catheterization was performed per pig, but all animals remained healthy without both any complications and hematological inflammatory abnormalities. The time course of neointimal proliferation of each stent was observed from the stage of hyperplasia to partial regression. The peak neointimal proliferation varied from 6 to 12 weeks despite implantation of identical stents using the same deployment method. In conclusion, repeated carotid artery catheterization to the same animal is feasible without animal health deterioration. This model should be useful to evaluate the time course of neointimal proliferation after stent deployment in preclinical study.