MilliporeSigma
  • Fluorescence Quenching-Based Mechanism for Determination of Hypochlorite by Coumarin-Derived Sensors.
Fluorescence Quenching-Based Mechanism for Determination of Hypochlorite by Coumarin-Derived Sensors.
International journal of molecular sciences (2019-01-16)
Karolina Starzak, Arkadiusz Matwijczuk, Bernadette Creaven, Alicja Matwijczuk, Sławomir Wybraniec, Dariusz Karcz
ABSTRACT
A fluorescence quenching-based mechanism for the determination of hypochlorite was proposed based on spectroscopic and chromatographic studies on the hypochlorite-sensing potency of three structurally similar and highly fluorescent coumarins. The mode of action was found to rely upon a chlorination of the coumarin-based probes resulting from their reaction with sodium hypochlorite. Importantly, the formation of chlorinated derivatives was accompanied by a linear decrease in the fluorescence intensities of the probes tested. The results obtained suggest the applicability of a coumarin-dependent hypochlorite recognition mechanism for the detection of, as well as for quantitative determination of, hypochlorite species in vitro.