MilliporeSigma
  • Anti-Thrombin, Anti-Adhesive, Anti-Migratory, and Anti-Proliferative Activities of Sulfated Galactans from the Tropical Green Seaweed, Udotea flabellum.
Anti-Thrombin, Anti-Adhesive, Anti-Migratory, and Anti-Proliferative Activities of Sulfated Galactans from the Tropical Green Seaweed, Udotea flabellum.
Marine drugs (2018-12-24)
Maxsuell Lucas Mendes Marques, Fernando Bastos Presa, Rony Lucas Silva Viana, Mariana Santana Santos Pereira Costa, Monica Oliveira Rocha Amorim, Daniel Lima Bellan, Monique Gabriela Chagas Faustino Alves, Leandro Silva Costa, Edvaldo Silva Trindade, Hugo Alexandre Oliveira Rocha
ABSTRACT
In this study, sulfated polysaccharide-rich extracts were isolated from 22 tropical seaweeds (4 red, 11 brown, and 7 green) found in northeastern Brazil, and evaluated for the role of anticoagulant agents. Fifteen of the extracts showed anticoagulant activity, including all the extracts from green seaweeds. Udoteaflabellum (a green seaweed) extract was the most potent, requiring an amount of only 3 µg to double the plasma coagulation time in the activated partial thromboplastin time test. A similar result was obtained with 1 µg of heparin. Two sulfated homogalactans with anticoagulant activity, F-I (130 kDa) and F-II (75 kDa), were isolated from this extract using several bio-guided purification steps. Their anticoagulant activity, as well as properties related to antitumor activity (anti-proliferative, anti-adhesive, and anti-migratory), were accessed. Their anticoagulant activities were close to that of heparin. We found that F-I and F-II (0.5⁻10 μg/mL) were not able to directly inhibit thrombin. In the presence of anti-thrombin, F-I (0.5 μg/mL) was more effective than heparin (0.5 μg/mL) in inhibiting thrombin, while F-II showed similar effects as heparin. F-I and F-II also inhibited B16-F10 (murine melanoma cells) adhesion, migration, and proliferation on a fibronectin-coated surface, but not on laminin- or collagen I-coated surfaces. Except for the antiproliferative activity, the other effects of F-I and F-II were eliminated upon their desulfation (~50%), indicating that the degree of sulfation is not as important for F-I and F-II anti-proliferative activity as the sulfation position. Taken together, the results provide strong evidence for the potential utility of sulfated galactans from U.flabellum, making these compounds an interesting option for future investigations that aim to design new anticoagulant/antitumor agents.