MilliporeSigma
  • Hypoxia-induced cancer stemness acquisition is associated with CXCR4 activation by its aberrant promoter demethylation.
Hypoxia-induced cancer stemness acquisition is associated with CXCR4 activation by its aberrant promoter demethylation.
BMC cancer (2019-02-15)
Nahyeon Kang, Su Yeon Choi, Bit Na Kim, Chang Dong Yeo, Chan Kwon Park, Young Kyoon Kim, Tae-Jung Kim, Seong-Beom Lee, Sug Hyung Lee, Jong Y Park, Mi Sun Park, Hyeon Woo Yim, Seung Joon Kim
ABSTRACT
A hypoxic microenvironment leads to an increase in the invasiveness and the metastatic potential of cancer cells within tumors via the epithelial-mesenchymal transition (EMT) and cancer stemness acquisition. However, hypoxia-induced changes in the expression and function of candidate stem cell markers and their possible molecular mechanism is still not understood. Lung cell lines were analyzed in normoxic or hypoxic conditions. For screening among the stem cell markers, a transcriptome analysis using next-generation sequencing was performed. For validation, the EMT and stem cell characteristics were analyzed. To determine whether an epigenetic mechanism was involved, the cell lines were treated with a DNA methyltransferase inhibitor (AZA), and methylation-specific PCR and bisulfite sequencing were performed. Next-generation sequencing revealed that the CXCR4 expression was significantly higher after the hypoxic condition, which functionally resulted in the EMT and cancer stemness acquisition. The acquisition of the EMT and stemness properties was inhibited by treatment with CXCR4 siRNA. The CXCR4 was activated by either the hypoxic condition or treatment with AZA. The methylation-specific PCR and bisulfite sequencing displayed a decreased CXCR4 promoter methylation in the hypoxic condition. These results suggest that hypoxia-induced acquisition of cancer stem cell characteristics was associated with CXCR4 activation by its aberrant promoter demethylation.