MilliporeSigma
  • Bimatoprost loaded nanovesicular long-acting sub-conjunctival in-situ gelling implant: In vitro and in vivo evaluation.
Bimatoprost loaded nanovesicular long-acting sub-conjunctival in-situ gelling implant: In vitro and in vivo evaluation.
Materials science & engineering. C, Materials for biological applications (2019-07-28)
Monika Yadav, Ana Guzman-Aranguez, Maria J Perez de Lara, Mandeep Singh, Joga Singh, Indu Pal Kaur
ABSTRACT
Primary treatment for glaucoma relies on chronic instillation (daily) of intraocular pressure (IOP) lowering eye drops. Present study tends to develop and assess a novel sustained release bimatoprost loaded nanovesicular (BMT-NV) - thermosensitive in-situ gelling implant (BMT-NV-GEL-IM), for subconjunctival delivery. BMT-NVs developed using novel composition and method of preparation, (IPA/700/DEL/2014) and industrially viable methodology were characterized and evaluated comprehensively for ocular suitability. Their incorporation into an in-situ gelling formula was safe (in vitro and in vivo) and stable upon sterilization. Autoclavability was an important consideration, as a preservative-free, single-use BMT-NV-GEL-IM will avoid side- effects associated with repetitive application of drops containing preservatives like benzalkonium chloride (BAK). An extended in vitro release of BMT (80.23%) was observed for 10 days while the IOP lowering effect extended over 2 months with single subconjunctival injection of BMT-NV-GEL-IM in rats. No clinical signs of irritation, inflammation, or infection were observed in any injected eye, throughout the study, as also confirmed by histology. Furthermore, single administration of BMT-NV-GEL as topical drop lowered the IOP over 5 days. Presence of significant diffuse fluorescence in confocal microscopy of internal eye tissues post-in vivo application, as subconjunctival implant, even after 2 month and eye drops upto1 week provide direct evidence of successful sustained delivery. We thus provide an improved modality for antiglaucoma medication in patients who are challenged to adhere to a regimen of daily eye drops.