MilliporeSigma
  • An Electrochemical Sandwich Immunosensor for the Detection of HER2 using Antibody-Conjugated PbS Quantum Dot as a label.
An Electrochemical Sandwich Immunosensor for the Detection of HER2 using Antibody-Conjugated PbS Quantum Dot as a label.
Journal of pharmaceutical and biomedical analysis (2019-07-03)
Zur Mira Azizah Nor Haiza Lah, Shahrul Ainliah Alang Ahmad, Muhammad Safwan Zaini, Mazliana Ahmad Kamarudin
ABSTRACT
A facile electrochemical sandwich immunosensor for the detection of a breast cancer biomarker, the human epidermal growth factor receptor 2 (HER2), was designed, using lead sulfide quantum dots-conjugated secondary HER2 antibody (Ab2-PbS QDs) as a label. Using Ab2-PbS QDs in the development of electrochemical immunoassays leads to many advantages such as straightforward synthesis and well-defined stripping signal of Pb(II) through acid dissolution, which in turn yields better sensing performance for the sandwiched immunosensor. In the bioconjugation of PbS QDs, the available amine and hydroxyl groups from secondary anti-HER2 and capped PbS QDs were bound covalently together via carbonyldiimidazole (CDI) acting as a linker. In order to quantify the biomarker, SWV signal was obtained, where the Pb2+ ions after acid dissolution in HCl was detected. The plated mercury film SPCE was also detected in situ. Under optimal conditions, HER2 was detected in a linear range from 1-100 ng/mL with a limit of detection of 0.28 ng/mL. The measures of satisfactory recoveries were 91.3% to 104.3% for the spiked samples, displaying high selectivity. Therefore, this method can be applied to determine HER2 in human serum.