MilliporeSigma
  • Decreased expression of ubiquilin‑1 following neonatal hypoxia‑ischemic brain injury in mice.
Decreased expression of ubiquilin‑1 following neonatal hypoxia‑ischemic brain injury in mice.
Molecular medicine reports (2019-05-07)
Li Luo, Yilin Liu, Xing Tu, Xuxin Ren, Wenyan Zhao, Jing Liu, Li Zhang, Weiqiang Chen, Pei Zhang, Weicai Wang, Lanhai Lü, Mengxia Wang
ABSTRACT
Ubiquilin‑1 (Ubqln), a ubiquitin‑like protein, regulates degradation of misfolded proteins and has been reported to have a crucial role in multiple pathologic and physiologic conditions. The current study was undertaken to investigate the expression of Ubqln in the brain of a neonatal hypoxia‑ischemic (HI) brain injury model induced using the Rice method with some modifications. Mouse pups at postnatal day 7 day were used in this study. Pups underwent permanent ligation of the left common carotid artery and a consecutive hypoxic challenge (8% O2 and 92% N2 for 120 min). The expression of Ubqln in the brain of pups following HI was analyzed by immunofluorescence staining and western blot analysis. Immunofluorescence staining demonstrated that Ubqln was extensively distributed in the cerebral cortex and hippocampus, and Ubqln was expressed in neurons, astrocytes and microglia in the brains of the HI brain injury model mice. Western blot analyses revealed decreased expression of Ubqln in the HI penumbra of the mouse model compared with Ubqln in the sham control group. The results of this study revealed that HI alters the expression of Ubqln, thus may provide a novel understanding of role of Ubqln in neonatal hypoxic ischemic encephalopathy.
Sigma-Aldrich
Anti-NeuN antibody produced in rabbit, affinity isolated antibody