MilliporeSigma
  • Nisin, a food preservative produced by Lactococcus lactis, affects the localization pattern of intermediate filament protein in HaCaT cells.
Nisin, a food preservative produced by Lactococcus lactis, affects the localization pattern of intermediate filament protein in HaCaT cells.
Anatomical science international (2018-10-26)
Norio Kitagawa, Takahito Otani, Tetsuichiro Inai
ABSTRACT
Nisin is a food preservative produced by Lactococcus lactis subsp. lactis. Previous blood biochemical research revealed that nisin has physiological effects in mammals; although the site of action has yet to be identified, keratinocytes have been proposed as a possible target. In this study, we investigated whether nisin affects keratinocytes by examining the effects on eukaryotic intermediate filaments in HaCaT human keratinocytes. Treatment with 93 μg/ml nisin for 24 h decreased the localization of the intermediate filament proteins cytokeratin (CK)5 and CK17 at the cell periphery, which were distributed in a limited area in a ring- or net-like shape. However, this was not observed upon treatment for 6 h. The results of a serial dilution assay revealed that the effect on CK17 localization depends on the nisin concentration and were observed at ≥47 μg/ml. Moreover, this effect was partially blocked by treatment with the calcium channel blocker bepridil. Thus, despite the long history of nisin as being safe for humans, it has measurable effects on the keratinocyte cytoskeleton. Our findings also indicate that CK5 and CK17 can serve as markers for evaluating the effects of nisin on keratinocytes.
Sigma-Aldrich
Anti-Keratin 5 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Keratin 17 antibody produced in rabbit, affinity isolated antibody