MilliporeSigma
  • Cholesterol-pyrene as a probe for cholesterol distribution on ordered and disordered membranes: Determination of spectral wavelengths.
Cholesterol-pyrene as a probe for cholesterol distribution on ordered and disordered membranes: Determination of spectral wavelengths.
PloS one (2018-08-11)
Claudia Almeida, Anaëlle De Wreede, Antonin Lamazière, Jesus Ayala-Sanmartin
ABSTRACT
Biological membranes contain a large variety of lipids species compartmentalized in different domains heterogeneous in size, composition and dynamics. Cholesterol induces membrane ordered domains thanks to its affinity for saturated lipids. Membrane domains had been studied with fluorescent probes either linked to phospholipids and proteins or as individual fluorophore. However, no efficient formulation of a cholesterol probe has been available so far. Herein, we described a cholesterol-pyrene probe behaviour in heterogeneous membranes. We characterised the pyrene fluorescence spectra in liquid-ordered (Lo) and liquid-disordered (Ld) membranes. Using statistical multivariate analysis, we found out the most appropriate wavelengths for membrane domains studies. 373 nm and 379 nm were the most discriminant wavelengths to follow the liquid-ordered and the liquid-disordered environments. Cholesterol clustering behaviour was quantified by the modulation of the cholesterol-pyrene excimers peak (474 nm). In liquid-ordered membranes at low temperature, cholesterol-pyrene was found as multimers and as monomers. At high temperature, the liquid-ordered status of the membrane decreases and cholesterol-pyrene tends to cluster. In liquid-disordered membranes, cholesterol-pyrene was present mostly as monomers and the small quantity of excimers increased with temperature. Cholesterol-pyrene was used to test the ceramide effect on membranes, and presented a behaviour in agreement with the cholesterol behaviour reported in the literature. Overall, the presented data show that cholesterol-pyrene is an efficient sensor to study liquid ordered and liquid disordered organisation in membranes.