MilliporeSigma
  • Discovery of glycerol phosphate modification on streptococcal rhamnose polysaccharides.
Discovery of glycerol phosphate modification on streptococcal rhamnose polysaccharides.
Nature chemical biology (2019-04-03)
Rebecca J Edgar, Vincent P van Hensbergen, Alessandro Ruda, Andrew G Turner, Pan Deng, Yoann Le Breton, Najib M El-Sayed, Ashton T Belew, Kevin S McIver, Alastair G McEwan, Andrew J Morris, Gérard Lambeau, Mark J Walker, Jeffrey S Rush, Konstantin V Korotkov, Göran Widmalm, Nina M van Sorge, Natalia Korotkova
ABSTRACT
Cell wall glycopolymers on the surface of Gram-positive bacteria are fundamental to bacterial physiology and infection biology. Here we identify gacH, a gene in the Streptococcus pyogenes group A carbohydrate (GAC) biosynthetic cluster, in two independent transposon library screens for its ability to confer resistance to zinc and susceptibility to the bactericidal enzyme human group IIA-secreted phospholipase A2. Subsequent structural and phylogenetic analysis of the GacH extracellular domain revealed that GacH represents an alternative class of glycerol phosphate transferase. We detected the presence of glycerol phosphate in the GAC, as well as the serotype c carbohydrate from Streptococcus mutans, which depended on the presence of the respective gacH homologs. Finally, nuclear magnetic resonance analysis of GAC confirmed that glycerol phosphate is attached to approximately 25% of the GAC N-acetylglucosamine side-chains at the C6 hydroxyl group. This previously unrecognized structural modification impacts host-pathogen interaction and has implications for vaccine design.