MilliporeSigma
  • Inhibition of Hsp70 Suppresses Neuronal Hyperexcitability and Attenuates Epilepsy by Enhancing A-Type Potassium Current.
Inhibition of Hsp70 Suppresses Neuronal Hyperexcitability and Attenuates Epilepsy by Enhancing A-Type Potassium Current.
Cell reports (2019-01-04)
Fang Hu, Jingheng Zhou, Yanxin Lu, Lizhao Guan, Ning-Ning Wei, Yi-Quan Tang, KeWei Wang
ABSTRACT
The heat shock protein 70 (Hsp70) is upregulated in response to stress and has been implicated as a stress marker in temporal lobe epilepsy (TLE). However, whether Hsp70 plays a pathologic or protective role in TLE remains unclear. Here we report a deleterious role of Hsp70 in kainic acid (KA)-induced seizures. Hsp70 expression is upregulated in a KA model of TLE, and silencing or inhibition of Hsp70 suppresses neuronal hyperexcitability and attenuates acute or chronic epilepsy by enhancing A-type potassium current in hippocampal neurons. Hsp70 upregulation leads to proteosomal degradation of Kv4-KChIP4a channel complexes primarily encoding neuronal A-type current. Furthermore, Hsp70 directly binds to the N terminus of auxiliary KChIP4a and targets Kv4-KChIP4a complexes to proteasome. Taken together, our findings reveal a role of Hsp70 in the pathogenesis of epilepsy through degradation of Kv4-KChIP4a complexes, and pharmacological inhibition of Hsp70 may represent therapeutic potential for epilepsy or hyperexcitability-related neurological disorders.
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Pifithrin-μ, ≥97% (HPLC), solid
Sigma-Aldrich
VER-155008, ≥98% (HPLC)