MilliporeSigma
  • Ovarian tumor domain-containing ubiquitin aldehyde binding protein 1 inhibits inflammation by regulating Nur77 stability.
Ovarian tumor domain-containing ubiquitin aldehyde binding protein 1 inhibits inflammation by regulating Nur77 stability.
Cellular signalling (2019-03-25)
Han Zhong Pei, Bin Huang, Hyeun-Wook Chang, Suk-Hwan Baek
ABSTRACT
Nur77 (NR4A1) plays an important role in various inflammatory responses. Nur77 is rapidly degraded in cells and its protein level is critically controlled. Although few E3 ligases regulating the Nur77 protein have been defined, the deubiquitinase (DUB) responsible for Nur77 stability has not been reported to date. We identified ovarian tumor domain-containing ubiquitin aldehyde binding protein 1 (OTUB1) as a DUB that stabilizes Nur77 by preventing its proteasomal degradation. We found that OTUB1 interacted with Nur77 to deubiquitinate it, thereby stabilizing Nur77 in an Asp88-dependent manner. This suggests that OTUB1 targets Nur77 for deubiquitination via a non-canonical mechanism. Functionally, OTUB1 inhibited TNFα-induced IL-6 production by promoting Nur77 protein stability. OTUB1 modulated the stability of Nur77 as a counterpart of tripartite motif 13 (Trim13). That is, OTUB1 reduced the ubiquitination and degradation of Nur77 potentiated by Trim13. In addition, this DUB also inhibited IL-6 production, which was further amplified by Trim13 in TNFα-induced responses. These findings suggest that OTUB1 is an important regulator of Nur77 stability and plays a role in controlling the inflammatory response.