MilliporeSigma
  • Novel dual-action prodrug triggers apoptosis in glioblastoma cells by releasing a glutathione quencher and lysine-specific histone demethylase 1A inhibitor.
Novel dual-action prodrug triggers apoptosis in glioblastoma cells by releasing a glutathione quencher and lysine-specific histone demethylase 1A inhibitor.
Journal of neurochemistry (2018-12-29)
Martin Engel, Yi Sing Gee, Dale Cross, Alan Maccarone, Benjamin Heng, Amy Hulme, Grady Smith, Gilles J Guillemin, Brett W Stringer, Christopher J T Hyland, Lezanne Ooi
ABSTRACT
Targeting epigenetic mechanisms has shown promise against several cancers but has so far been unsuccessful against glioblastoma (GBM). Altered histone 3 lysine 4 methylation and increased lysine-specific histone demethylase 1A (LSD1) expression in GBM tumours nonetheless suggest that epigenetic mechanisms are involved in GBM. We engineered a dual-action prodrug, which is activated by the high hydrogen peroxide levels associated with GBM cells. This quinone methide phenylaminecyclopropane prodrug releases the LSD1 inhibitor 2-phenylcyclopropylamine with the glutathione scavenger para-quinone methide to trigger apoptosis in GBM cells. Quinone methide phenylaminocyclopropane impaired GBM cell behaviours in two-dimensional and three-dimensional assays, and triggered cell apoptosis in several primary and immortal GBM cell cultures. These results support our double-hit hypothesis of potentially targeting LSD1 and quenching glutathione, in order to impair and kill GBM cells but not healthy astrocytes. Our data suggest this strategy is effective at selectively targeting GBM and potentially other types of cancers. OPEN SCIENCE BADGES: This article has received a badge for *Open Materials* because it provided all relevant information to reproduce the study in the manuscript. The complete Open Science Disclosure form for this article can be found at the end of the article. More information about the Open Practices badges can be found at https://cos.io/our-services/open-science-badges/.
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, (H+L) HRP conjugate, 1 mg/mL, Chemicon®
Sigma-Aldrich
trans-2-Phenylcyclopropylamine hydrochloride
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, serum, Upstate®
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody