MilliporeSigma
  • Altered in Vitro Metabolomic Response of the Human Microbiota to Sweeteners.
Altered in Vitro Metabolomic Response of the Human Microbiota to Sweeteners.
Genes (2019-07-18)
Emanuel Vamanu, Diana Pelinescu, Florentina Gatea, Ionela Sârbu
ABSTRACT
Non-nutritive sweeteners represent an ingredient class that directly affects human health, via the development of inflammatory processes that promote chronic diseases related to microbiota dysbiosis. Several in vitro tests were conducted in the static GIS1 simulator. The aim of the study was to highlight the effect of sweeteners on the microbiota pattern of healthy individuals, associated with any alteration in the metabolomic response, through the production of organic acids and ammonium. The immediate effect of the in vitro treatment and the influence of the specific sweetener type on the occurrence of dysbiosis were evaluated by determining the biomarkers of the microbiota response. The presence of the steviol reduced the ammonium level (minimum of 410 mg/L), while the addition of cyclamate and saccharin caused a decrease in the number of microorganisms, in addition to lowering the total quantity of synthesized short-chain fatty acids (SCFAs). The bifidobacteria appeared to decrease below 102 genomes/mL in all the analyzed samples at the end of the in vitro simulation period. Barring the in vitro treatment of steviol, all the sweeteners tested exerted a negative influence on the fermentative profile, resulting in a decline in the fermentative processes, a rise in the colonic pH, and uniformity of the SCFA ratio.