MilliporeSigma
  • Loss of the transcription factor RBPJ induces disease-promoting properties in brain pericytes.
Loss of the transcription factor RBPJ induces disease-promoting properties in brain pericytes.
Nature communications (2019-06-30)
Rodrigo Diéguez-Hurtado, Katsuhiro Kato, Benedetto Daniele Giaimo, Melina Nieminen-Kelhä, Hendrik Arf, Francesca Ferrante, Marek Bartkuhn, Tobias Zimmermann, M Gabriele Bixel, Hanna M Eilken, Susanne Adams, Tilman Borggrefe, Peter Vajkoczy, Ralf H Adams
ABSTRACT
Sufficient vascular supply is indispensable for brain development and function, whereas dysfunctional blood vessels are associated with human diseases such as vascular malformations, stroke or neurodegeneration. Pericytes are capillary-associated mesenchymal cells that limit vascular permeability and protect the brain by preserving blood-brain barrier integrity. Loss of pericytes has been linked to neurodegenerative changes in genetically modified mice. Here, we report that postnatal inactivation of the Rbpj gene, encoding the transcription factor RBPJ, leads to alteration of cell identity markers in brain pericytes, increases local TGFβ signalling, and triggers profound changes in endothelial behaviour. These changes, which are not mimicked by pericyte ablation, imperil vascular stability and induce the acquisition of pathological landmarks associated with cerebral cavernous malformations. In adult mice, loss of Rbpj results in bigger stroke lesions upon ischemic insult. We propose that brain pericytes can acquire deleterious properties that actively enhance vascular lesion formation and promote pathogenic processes.
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - Cy3 antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-AQP4 antibody produced in rabbit, affinity isolated antibody, Prestige Antibodies® Powered by Atlas Antibodies, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-NG2 Chondroitin Sulfate Proteoglycan Antibody, Chemicon®, from rabbit