MilliporeSigma
A protein quality control pathway regulated by linear ubiquitination.
The EMBO journal (2019-03-20)
Eva M van Well, Verian Bader, Maria Patra, Ana Sánchez-Vicente, Jens Meschede, Nikolas Furthmann, Cathrin Schnack, Alina Blusch, Joseph Longworth, Elisabeth Petrasch-Parwez, Kohji Mori, Thomas Arzberger, Dietrich Trümbach, Lena Angersbach, Cathrin Showkat, Dominik A Sehr, Lena A Berlemann, Petra Goldmann, Albrecht M Clement, Christian Behl, Andreas C Woerner, Carsten Saft, Wolfgang Wurst, Christian Haass, Gisa Ellrichmann, Ralf Gold, Gunnar Dittmar, Mark S Hipp, F Ulrich Hartl, Jörg Tatzelt, Konstanze F Winklhofer
ABSTRACT
Neurodegenerative diseases are characterized by the accumulation of misfolded proteins in the brain. Insights into protein quality control mechanisms to prevent neuronal dysfunction and cell death are crucial in developing causal therapies. Here, we report that various disease-associated protein aggregates are modified by the linear ubiquitin chain assembly complex (LUBAC). HOIP, the catalytic component of LUBAC, is recruited to misfolded Huntingtin in a p97/VCP-dependent manner, resulting in the assembly of linear polyubiquitin. As a consequence, the interactive surface of misfolded Huntingtin species is shielded from unwanted interactions, for example with the low complexity sequence domain-containing transcription factor Sp1, and proteasomal degradation of misfolded Huntingtin is facilitated. Notably, all three core LUBAC components are transcriptionally regulated by Sp1, linking defective LUBAC expression to Huntington's disease. In support of a protective activity of linear ubiquitination, silencing of OTULIN, a deubiquitinase with unique specificity for linear polyubiquitin, decreases proteotoxicity, whereas silencing of HOIP has the opposite effect. These findings identify linear ubiquitination as a protein quality control mechanism and hence a novel target for disease-modifying strategies in proteinopathies.
Sigma-Aldrich
Anti-OPTN antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-HNRNPL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Huntingtin Protein Antibody, clone mEM48, culture supernatant, clone mEM48, Chemicon®
Sigma-Aldrich
Anti-Huntingtin Protein Antibody, a.a. 181-810, clone 1HU-4C8, ascites fluid, clone 1HU-4C8, Chemicon®
Sigma-Aldrich
Anti-HA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
(R)-MG132
Sigma-Aldrich
NMS-873, ≥98% (HPLC)